siRNA Details
Gene Symbol: AGBL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF075049.1 | - | 1 | 130 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | H38784.1 | - | 1 | 130 | Chr. 1 - 49690436 - 49690829 [-] on Build GRCh38 |
