siRNA Details
Gene Symbol: ZNF814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC026064.2 | - | 1 | 355 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE796049.1 | - | 1 | 335 | Chr. 19 - 57885923 - 57889493 [-] on Build GRCh38 |
