siRNA Details
Gene Symbol: LOC388406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126009.1 | 3, 4 | 339 | |||
GenBank mRNA | AK131012.1 | - | 3, 4 | 340 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC388406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126009.1 | 3, 4 | 339 | |||
GenBank mRNA | AK131012.1 | - | 3, 4 | 340 |