siRNA Details
Gene Symbol: LINC01093 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039975.1 | 3 | 1353 | |||
NR_039976.1 | 3 | 1340 | ||||
NR_039977.1 | 2 | 1120 | ||||
GenBank mRNA | AK096134.1 | - | 3 | 2873 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01093 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039975.1 | 3 | 1353 | |||
NR_039976.1 | 3 | 1340 | ||||
NR_039977.1 | 2 | 1120 | ||||
GenBank mRNA | AK096134.1 | - | 3 | 2873 |