siRNA Details
Gene Symbol: LOC338694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104161.1 | 3 | 665 | |||
GenBank mRNA | BC043531.1 | - | 3 | 665 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI826779.1 | - | 3 | 667 | Chr. 11 - 69147386 - 69171564 [-] on Build GRCh38 |
