siRNA Details
Gene Symbol: LOC101927780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110416.1 | 4 | 441 | |||
NR_110417.1 | 3 | 177 | ||||
GenBank mRNA | BX648502.1 | - | 4 | 443 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY029810.1 | - | 4 | 398 | Chr. 14 - 61565120 - 61586222 [-] on Build GRCh38 |
