siRNA Details
Gene Symbol: STMN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131326.1 | - | 1 | 131 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR613563.1 | - | 1 | 155 | Chr. 20 - 63639845 - 63641845 [-] on Build GRCh38 |
