siRNA Details
Gene Symbol: TTC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017017950.1 | XP_016873439.1 | 24 | 2370 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK027161.1 | - | 1 | 1674 | Chr. 11 - 113369810 - 113371541 [+] on Build GRCh38 |
