siRNA Details
Gene Symbol: LOC101927494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135563.1 | 2 | 586 | |||
GenBank mRNA | BC035252.1 | - | 2 | 586 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI545414.1 | - | 2 | 591 | Chr. 3 - 86481943 - 86496439 [+] on Build GRCh38 |
