siRNA Details
Gene Symbol: LOC730202 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC019017.2 | - | 1 | 728 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE902834.1 | - | 1 | 706 | Chr. 14 - 96501465 - 96502301 [-] on Build GRCh38 |
