siRNA Details
Gene Symbol: LOC101928150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120634.1 | 2 | 233 | |||
GenBank mRNA | CR749724.1 | - | 2 | 233 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL044810.1 | - | 2 | 233 | Chr. 10 - 6618716 - 6625346 [+] on Build GRCh38 |
