siRNA Details
Gene Symbol: CDK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC001274.1 | - | 9 | 1393 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK024379.1 | - | 1 | 2662 | Chr. 7 - 40066704 - 40070406 [+] on Build GRCh38 |
