siRNA Details
Gene Symbol: LOC101928650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125958.1 | 2 | 243 | |||
GenBank mRNA | AK096329.1 | - | 3 | 241 | ||
BC065239.1 | - | 2 | 341 | |||
HY028703.1 | - | 3 | 344 | |||
HY035829.1 | - | 2 | 243 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU161108.1 | - | 2 | 338 | Chr. 1 - 170461936 - 170494533 [-] on Build GRCh38 |
