siRNA Details
Gene Symbol: LOC101928650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC065239.1 | - | 1 | 150 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU161108.1 | - | 1 | 147 | Chr. 1 - 170461936 - 170494533 [-] on Build GRCh38 |
