siRNA Details
Gene Symbol: LOC101928460 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110758.1 | 1 | 78 | |||
GenBank mRNA | BC040893.1 | - | 1 | 78 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM554569.1 | - | 1 | 103 | Chr. 1 - 29330735 - 29350114 [-] on Build GRCh38 |
