siRNA Details
Gene Symbol: CLDN10-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046533.1 | 3 | 356 | |||
GenBank mRNA | BC024972.1 | - | 3 | 357 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG574725.1 | - | 3 | 354 | Chr. 13 - 95479522 - 95533902 [-] on Build GRCh38 |
