siRNA Details
Gene Symbol: LOC440934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136642.1 | 4 | 3920 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK055286.1 | - | 1 | 907 | Chr. 2 - 222355483 - 222356726 [+] on Build GRCh38 |
