siRNA Details
Gene Symbol: LOC101927701 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110271.1 | 3 | 629 | |||
GenBank mRNA | BC028245.1 | - | 3 | 640 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI861426.1 | - | 3 | 625 | Chr. 2 - 67796054 - 67825561 [-] on Build GRCh38 |
