siRNA Details
Gene Symbol: LOC101927697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109888.1 | 6 | 680 | |||
GenBank mRNA | BC047601.1 | - | 6 | 680 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM804615.1 | - | 6 | 681 | Chr. 5 - 158323455 - 158409772 [-] on Build GRCh38 |
