siRNA Details
Gene Symbol: SAMHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005260384.3 | XP_005260441.1 | 15 | 3659 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SAMHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005260384.3 | XP_005260441.1 | 15 | 3659 |