siRNA Details
Gene Symbol: LOC101927919 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110854.1 | 1 | 105 | |||
GenBank mRNA | AK125564.1 | - | 1 | 105 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA896292.1 | - | 1 | 105 | Chr. 6 - 117673953 - 117675284 [-] on Build GRCh38 |
