siRNA Details
Gene Symbol: LINC00847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095170.1 | - | 1 | 58 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA351348.1 | - | 1 | 58 | Chr. 5 - 180832537 - 180833128 [+] on Build GRCh38 |
