siRNA Details
Gene Symbol: LINC00847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045679.1 | 2 | 410 | |||
GenBank mRNA | AK095170.1 | - | 1 | 273 | ||
BC047586.1 | - | 2 | 342 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045679.1 | 2 | 410 | |||
GenBank mRNA | AK095170.1 | - | 1 | 273 | ||
BC047586.1 | - | 2 | 342 |