siRNA Details
Gene Symbol: GAPLINC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110428.1 | 3 | 398 | |||
NR_110429.1 | 3 | 424 | ||||
GenBank mRNA | AJ707365.1 | - | 3 | 361 | ||
BM782281.1 | - | 3 | 398 | |||
BX107736.1 | - | 3 | 336 |
