siRNA Details
Gene Symbol: CAMKK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128601.1 | - | 8 | 2840 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK126617.1 | - | 1 | 837 | Chr. 17 - 3878826 - 3880725 [-] on Build GRCh38 |
