siRNA Details
Gene Symbol: LOC101928205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110827.1 | 2 | 563 | |||
GenBank mRNA | BC034818.1 | - | 2 | 563 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG724391.1 | - | 2 | 569 | Chr. 17 - 72072333 - 72082063 [+] on Build GRCh38 |
