siRNA Details
Gene Symbol: COL27A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032888.3 | NP_116277.2 | 61 | 7217 | ||
XM_006717308.2 | XP_006717371.1 | 60 | 7231 | |||
XM_006717310.3 | XP_006717373.1 | 59 | 5030 | |||
XM_011519138.2 | XP_011517440.1 | 61 | 6849 | |||
XM_011519140.1 | XP_011517442.1 | 60 | 7231 | |||
XM_011519145.2 | XP_011517447.1 | 48 | 4459 |
