siRNA Details
Gene Symbol: DNM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000979.1 | XP_016856468.1 | 23 | 2940 | ||
XM_017000981.1 | XP_016856470.1 | 21 | 2784 | |||
XM_017000983.1 | XP_016856472.1 | 21 | 2766 | |||
XM_017000984.1 | XP_016856473.1 | 20 | 2754 | |||
GenBank mRNA | AL035110.1 | - | 4 | 711 |
