siRNA Details
Gene Symbol: WDFY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011534914.1 | XP_011533216.1 | 12 | 5789 | ||
XM_011534915.2 | XP_011533217.1 | 11 | 8597 | |||
XM_017020386.1 | XP_016875875.1 | 12 | 14321 | |||
GenBank mRNA | AK095358.1 | - | 1 | 2907 |
