siRNA Details
Gene Symbol: RUSC1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125510.1 | - | 1 | 2602 | ||
BC131614.1 | - | 2 | 1060 | 40115930 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RUSC1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125510.1 | - | 1 | 2602 | ||
BC131614.1 | - | 2 | 1060 | 40115930 |