siRNA Details
Gene Symbol: BTG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC028229.1 | - | 1 | 1665 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX101397.1 | - | 1 | 83 | Chr. 21 - 17613355 - 17613859 [+] on Build GRCh38 |
