siRNA Details
Gene Symbol: LOC101929532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110255.1 | 4 | 3372 | |||
GenBank mRNA | BX648358.1 | - | 4 | 3369 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110255.1 | 4 | 3372 | |||
GenBank mRNA | BX648358.1 | - | 4 | 3369 |