siRNA Details
Gene Symbol: HOXB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M92299.1 | - | 1 | 78 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DN996030.1 | - | 1 | 110 | Chr. 17 - 48593447 - 48593985 [-] on Build GRCh38 |
