siRNA Details
Gene Symbol: LINC00667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC027873.1 | - | 1 | 1365 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BP417823.1 | - | 1 | 417 | Chr. 18 - 5238790 - 5239238 [+] on Build GRCh38 |
