siRNA Details
Gene Symbol: ALMS1-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046762.1 | 2 | 365 | |||
GenBank mRNA | BC041487.1 | - | 2 | 372 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM451564.1 | - | 371 | Chr. - - [-] on Build |
