siRNA Details
Gene Symbol: ABCB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011538098.2 | XP_011536400.1 | 1 | 1605 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF461897.1 | - | 1 | 652 | Chr. 12 - 122951241 - 122952311 [-] on Build GRCh38 |
