siRNA Details
Gene Symbol: C18orf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026109.1 | XP_016881598.1 | 1 | 382 | ||
GenBank mRNA | AB027121.1 | - | 1 | 382 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C18orf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026109.1 | XP_016881598.1 | 1 | 382 | ||
GenBank mRNA | AB027121.1 | - | 1 | 382 |