siRNA Details
Gene Symbol: TMPRSS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AW295507.1 | - | 1 | 292 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK055306.1 | - | 1 | 930 | Chr. 22 - 37064783 - 37067484 [+] on Build GRCh38 |
