siRNA Details
Gene Symbol: C7orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012589.1 | XP_016868078.1 | 4 | 5840 | ||
XM_017012590.1 | XP_016868079.1 | 4 | 5766 | |||
XM_017012591.1 | XP_016868080.1 | 4 | 5620 | |||
XM_017012592.1 | XP_016868081.1 | 3 | 5801 | |||
XM_017012593.1 | XP_016868082.1 | 4 | 5670 | |||
XM_017012594.1 | XP_016868083.1 | 4 | 5877 |
