siRNA Details
Gene Symbol: LOC101929260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110241.1 | 4 | 401 | |||
GenBank mRNA | BC014936.1 | - | 4 | 401 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE894820.1 | - | 4 | 380 | Chr. 2 - 150569578 - 150572200 [-] on Build GRCh38 |
