siRNA Details
Gene Symbol: LOC101929260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110241.1 | 5 | 785 | |||
GenBank mRNA | BC014936.1 | - | 5 | 785 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110241.1 | 5 | 785 | |||
GenBank mRNA | BC014936.1 | - | 5 | 785 |