siRNA Details
Gene Symbol: LOC401433 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127717.1 | - | 1 | 65 |
Gene Symbol: WDR86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516144.2 | XP_011514446.1 | 7 | 1772 | ||
XM_011516145.2 | XP_011514447.1 | 6 | 1643 | |||
XM_011516147.2 | XP_011514449.1 | 6 | 1572 | |||
XM_011516151.2 | XP_011514453.1 | 5 | 1443 |
