siRNA Details
Gene Symbol: LIPE-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073179.1 | 2 | 2247 | |||
NR_073180.1 | 3 | 1002 | ||||
GenBank mRNA | AK096849.1 | - | 2 | 2246 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LIPE-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073179.1 | 2 | 2247 | |||
NR_073180.1 | 3 | 1002 | ||||
GenBank mRNA | AK096849.1 | - | 2 | 2246 |