siRNA Details
Gene Symbol: LOC101927358 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121182.1 | 2, 3 | 536 | |||
GenBank mRNA | BC021182.1 | - | 2, 3 | 541 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG720148.1 | - | 2, 3 | 541 | Chr. 9 - 74371531 - 74383530 [+] on Build GRCh38 |
