siRNA Details
Gene Symbol: LRRC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017001886.1 | XP_016857375.1 | 30 | 8751 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC037323.1 | - | 1 | 118 | Chr. 1 - 70141382 - 70144364 [+] on Build GRCh38 |
