siRNA Details
Gene Symbol: PRMT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017008823.1 | XP_016864312.1 | 12 | 4563 | ||
XM_017008824.1 | XP_016864313.1 | 11 | 4495 | |||
XM_017008825.1 | XP_016864314.1 | 11 | 4549 | |||
XM_017008826.1 | XP_016864315.1 | 10 | 4444 |
