siRNA Details
Gene Symbol: AGO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017629.3 | NP_060099.2 | 18 | 6531 | ||
XM_005270578.3 | XP_005270635.1 | 18 | 6416 | |||
XM_005270579.3 | XP_005270636.1 | 17 | 6224 | |||
XM_011540883.2 | XP_011539185.1 | 15 | 6020 | |||
XM_011540884.2 | XP_011539186.1 | 15 | 5883 |
