siRNA Details
Gene Symbol: C18orf63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174123.1 | NP_001167594.1 | 14 | 3007 | ||
GenBank mRNA | CR749350.1 | - | 14 | 3007 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C18orf63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174123.1 | NP_001167594.1 | 14 | 3007 | ||
GenBank mRNA | CR749350.1 | - | 14 | 3007 |