siRNA Details
Gene Symbol: NAV2-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC018444.1 | - | 2 | 405 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG168093.1 | - | 2 | 404 | Chr. 11 - 19380485 - 19385014 [+] on Build GRCh38 |
