siRNA Details
Gene Symbol: ADAMTS9-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038264.1 | 2 | 611 | |||
GenBank mRNA | AK057923.1 | - | 2 | 379 | ||
AL832788.1 | - | 2 | 572 | |||
BC040632.1 | - | 2 | 353 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ADAMTS9-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038264.1 | 2 | 611 | |||
GenBank mRNA | AK057923.1 | - | 2 | 379 | ||
AL832788.1 | - | 2 | 572 | |||
BC040632.1 | - | 2 | 353 |